Learn More
Medchemexpress LLC Stat3-in-48 | 1313019-65-6 | 98.0% | 431.80 | C21H13ClF3N3O2 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1366585MG
Encompass_Preferred
STAT3-IN-48 is a small-molecule research compound that inhibits phosphorylation of STAT3 and induces apoptosis via SHP-1-dependent STAT3 inactivation. It is intended for use in cellular and biochemical studies of the JAK/STAT signaling pathway and cancer research.
  • Inhibits phosphorylation of STAT3.
  • Induces apoptosis through SHP-1-dependent STAT3 inactivation.
  • Structurally related to sorafenib.
  • Suitable for in vitro cellular and biochemical assays targeting the JAK/STAT pathway.
  • Reported HPLC purity 98.02% (per COA).
Catalog No. 50-002-21953