Learn More
Abcam STAT3 pY705 ELISA Kit, 1 x 96 Tests
Supplier:  Abcam AB1766541X96TESTS
Encompass_Preferred
STAT3 pY705 ELISA Kit is a single-wash 90-min Simplestep used to quantify STAT3 pY705 with a sensitivity of 30 µg/mL. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC1658014