Learn More
Cayman Chemical STAT5 InhIbItr 25mg
SDP
Supplier:  Cayman Chemical 1578425 MG
Encompass_Preferred
A cell-permeable, nonpetidic nicotinoyl hydrazone that selectively suppresses STAT5 by targeting its SH2 domain (IC50 = 47 µM against STAT5β SH2 domain EPO peptide binding activity)
Catalog No. 50-297-2720