Learn More
Medchemexpress LLC STAT6 Human His 100ug | 100ug
SDP
Supplier:  Medchemexpress LLC HYP71337100UG
Encompass_Preferred
STAT6 is a multifunctional protein that performs dual functions of signal transduction and transcriptional activation It plays a crucial role in mediating IL4/interleukin-4 and IL3/interleukin-3 induced signaling cascades STAT6 Protein Human (His) is the recombinant human-derived STAT6 protein expressed by E coli with C-6 His labeled tag
Catalog No. 50-001-96643