Learn More
Medchemexpress LLC STAT6 Human His 5ug

Supplier: Medchemexpress LLC HYP713375UG
Recombinant human STAT6 (His) is supplied lyophilized for research use in studies of IL-4/IL-13 signaling, signal transduction, and transcriptional regulation. The protein is expressed in E. coli and provided as a small, research-scale vial intended for biochemical assays and signaling studies. Follow storage and reconstitution recommendations to maintain protein integrity and activity.
- Contains C-terminal His tag for affinity purification or detection.
- Produced in E. coli expression system.
- Lyophilized formulation for long-term storage.
- High purity: >95% by reducing SDS-PAGE.
- Low endotoxin: <1 EU/μg (LAL method).
- Store at -20°C; reconstitute to ≥100 μg/mL recommended.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.