Learn More
MALIX LLC Status BTA (25 Tests)A Rapid Test For the Qualitative Detection of Bladder Tumor Associated Antigenin Human Urine
Supplier:  MALIX LLC STATBTA
Encompass_Preferred
Status BTA (25 Tests)
The test is indicated for use as an aid in the management of bladder cancer patients in conjunction with cystoscopy. The simple urine sample collection also allows for improved patient comfort. Because tumor markers can be used to assess the response of a tumor treatment and for prognosis, rapid screening tests like the Status BTA product are very helpful in the treatment management of cancer patients.
Key Features:
CLIA Waived
Results available in 5 minutes
Simple procedure requires only adding 3 drops of urine to test device; no sample preparation needed
Accurate test results with excellent sensitivity and specificity
Uses monoclonal antibodies to detect the presence of bladder tumor-associated antigen in urine
Detects early state recurrence cancer that cytology may miss
Adjunct test to cystoscopy
Catalog No. NC3235788