Learn More
MALIX LLC Status™ Flu A & B Test: Rapid qualitative detection of influenza A and B antigens from nasal and nasopharyngeal swabs. CLIA waived, 25 tests per kit, 15-minute results, detects H1N1 (Swine Flu), easy-to-read.
Supplier: MALIX LLC STACOV
Status COVID-19/Flu A&B
The Status™ COVID-19/Flu A&B Test is a rapid immunoassay designed for the simultaneous direct detection and differential diagnosis of SARS-CoV-2, influenza type A, and influenza type B antigens from anterior nasal and nasopharyngeal swab specimens.
COVID-19 Detection:
Anterior Nasal Swab Specimen: Sensitivity 93.8%, Specificity 100%
Nasopharyngeal Swab Specimen: Sensitivity 93.1%, Specificity 100%
Flu A Detection:
Sensitivity 91.4%, Specificity 95.7%
Flu B Detection:
Sensitivity 87.6%, Specificity 95.9%
The test has received FDA Emergency Use Authorization (EUA) and provides visually readable results in 15 minutes. It utilizes a flocked nasopharyngeal swab for superior specimen collection and patient comfort. This test is an efficient tool for the rapid diagnosis of multiple respiratory infections, ensuring timely and accurate patient management.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.