Learn More
MALIX LLC For The Qualitative Detection Of Infectious Mononucleosis Heterophile Antibodies In Whole Blood
Supplier:  MALIX LLC STATMON10
Encompass
Status Mono w/ Controls (10 Tests)
For The Qualitative Detection Of Infectious Mononucleosis Heterophile Antibodies In Whole Blood
CLIA WAIVED for whole blood
FDA cleared with no age restrictions
Single reagent procedure provides results in 3-8 minutes
Fingerstick whole blood convenience
Superior 99.9% sensitivity for earlier IM detection, specificity of 98.8%
Multiple kit sizes available, ideal for any practice
External QC negative and positive controls included (item # 68364 & 84M30)
New plastic sample transfer pipettes
Catalog No. NC3235644