Learn More
MALIX LLC For The Qualitative Detection Of Infectious Mononucleosis Heterophile Antibodies In Whole Blood, Serum Or Plasma
Supplier:  MALIX LLC STATMON30
Encompass
Status™ Mono w/ controls 30 tests
The Status Mono test is for the qualitative detection of infectious mononucleosis (IM) heterophile antibodies in whole blood, serum, or plasma. It is CLIA waived for whole blood and FDA cleared with no age restrictions. This single reagent procedure provides rapid results in 3-8 minutes, making it convenient for quick diagnosis. The test offers fingerstick whole blood convenience, with superior sensitivity of 99.9% for early IM detection and a specificity of 98.8%. Additionally, the kit includes new plastic sample transfer pipettes for easy handling.
Key Features:
Qualitative detection of IM heterophile antibodies
CLIA waived for whole blood
FDA cleared with no age restrictions
Results in 3-8 minutes
Fingerstick whole blood
Sensitivity: 99.9%, Specificity: 98.8%
Includes plastic sample transfer pipettes
Test ensures timely and accurate detection
Catalog No. NC3235634