Learn More
TARGETMOL CHEMICALS INC STAUROSPORINE 10MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T668010MG
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Staurosporine (AM-2282) is a potent PKC inhibitor for PKC(alpha)/(gamma)/(eta) (IC50 2/5/4 nM) less potent to PKC(epsilon) (73 nM) PKC(delta) (20 nM) and little action to PKC(zeta) (1086 nM). purity: 99%
Catalog No. 50-257-9308