Learn More
Selleck Chemical LLC Stavudine (d4T) 25mg 3056-17-5 BMY-27857, Sanilvudine, NSC 163661
SDP
Supplier:  Selleck Chemical LLC S139825MG
Encompass_Preferred
Stavudine (d4T, BMY-27857, Sanilvudine, NSC 163661) is a nucleoside analog reverse transcriptase inhibitor (NARTI) active against HIV. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-629-9