Learn More
Abcam StayGreen/AP Plus (Alcohol and x ylene Substitute Compatible), 1KIT
Supplier:  Abcam AB1564281KIT
Encompass_Preferred
StayGreen/AP is a substrate chromogen system designed to be used for either IHC or ISH when using alkaline phosphate detection.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-258-2853