Learn More
Perkin Elmer US LLC Interference Check Solution 1, 5% HNO3/trace HF, 125 mL
Supplier:  Perkin Elmer US LLC N9303828
Encompass_Preferred
Concentration of Element includes: Matrix Concentration 5% HNO3/trace HF 10,000 µg/mL: Cl 2,000 µg/mL: C 1,000 µg/mL: Al, Ca, Fe, K, Mg, Na, P, S 20 µg/mL: Mg, Ti
Catalog No. 50-904-7980