Learn More
Tosoh Bioscience LLC STD PEO SE-5 39000MW 0.5G
Supplier:  Tosoh Bioscience LLC 0006212
Encompass
Calibration Standard, Polyethylene Oxide; Tosoh; Description: SE-5; Molecular Weight: 3.9 x 104 M. W.; Quantity: 0.5g
Catalog No. 50-851-142