Learn More
MSC Proto Socket, 1/4 in. Drive, 3/8 in. 12-Point, 1 Each
Supplier:  MSC 39934062
Encompass
This Proto 12-point socket is engineered for use on stubborn, damaged, or soft material fasteners. It features the TorquePlus™ design, a high-performance drive configuration that ensures quick and easy engagement with fastener flats, minimizing slippage and distributing contact stresses over a larger area to prevent rounding and extend fastener life.
  • Drive size: 1/4 in.
  • Socket size: 3/8 in.
  • Features 12 points for enhanced fastener engagement
  • Standard length
  • Chrome finish for durability
  • Minimizes fastener rounding with TorquePlus™ design
  • Overall length: 7/8 in.
  • Product weight: 0.03 lbs
Catalog No. 50-192-0330