Learn More
World Precision Instrument Wire Mesh Sterilization Baskets, Drop Handles-WP-4408
SDP
Supplier:  World Precision Instrument WP4408
Encompass
Stainless steel, electropolished Wire Mesh Sterilization Baskets with drop handles for instrument cleaning, sterilization and storage. The baskets meet all requirements of automatic washing and sterilization for all types of instruments.
Catalog No. 50-209-0259