Learn More
Biotium CellBrite™ Steady 488 Membrane Staining Kit | 100 labelings
Supplier: Biotium 30106T
CellBrite™ Steady Membrane Staining Kits allow fluorescence imaging of cell surface for up to several days in culture. The CellBrite™ Steady Dyes are unique fluorescent membrane probes that distribute between the cell surface and intracellular compartments, so cells retain cell surface staining over time. With the use of CellBrite™ Steady Enhancer, intracellular staining can be reduced or eliminated for imaging of cell outlines or boundaries.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.