Learn More
OriGene STEAP3 CytoSection 25 Slides
SDP
Supplier:  OriGene TS424274P5
Encompass_Preferred
Transient overexpression of STEAP3, transcript variant 2, in HEK293T cells, FFPE control for IHC, ICC and ISH staining, 25 slides per pack
Catalog No. NC3734427