Learn More
Cayman Chemical StearIc AcId-13C 1g
SDP
Supplier:  Cayman Chemical 289171 G
Encompass_Preferred
An internal standard for the quantification of stearic acid by GC- or LC-MS
Catalog No. 50-294-1827