Learn More
Medchemexpress LLC Stearic acid-13C18 | 287100-83-8 | 98.0% | 1 MG
Supplier: Medchemexpress LLC HYB2219S21MG
Stearic acid-13C18 is the 13C-labeled Stearic acid, a long chain dietary saturated fatty acid found in many animal and vegetable fats and oils. It is intended for research use only and not for sale to patients.
- Can be used as a tracer
- Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS