Learn More
Medchemexpress LLC Stearic acid-d2 | 19905-58-9 | 99.1% | 50 MG
SDP
Supplier:  Medchemexpress LLC HYB2219S150MG
Encompass_Preferred
Stearic acid-d2 is the deuterium labeled Stearic acid. Stearic acid is a long chain dietary saturated fatty acid which exists in many animal and vegetable fats and oils.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process.
  • Deuteration has potential to affect the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-10105