Learn More
Cayman Chemical StearIc AcId-d35 100mg
SDP
Supplier:  Cayman Chemical 9003318100 MG
Encompass_Preferred
An internal standard for the quantification of stearic acid by GC- or LC-MS
Catalog No. 50-289-6084