Learn More
Medchemexpress LLC Stearic acid-d35 | 17660-51-4 | 99.9% | 100 MG
SDP
Supplier:  Medchemexpress LLC HYB2219S4100MG
Encompass_Preferred
Stearic acid-d35 is the deuterium labeled Stearic acid, a long chain dietary saturated fatty acid found in many animal and vegetable fats and oils. This compound is intended for research use only.
  • Used as a tracer
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-09719