Learn More
Medchemexpress LLC Stearic acid-d35 | 17660-51-4 | 99.9% | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYB2219S4
Encompass_Preferred
Stearic acid-d35 is the deuterium labeled Stearic acid, a long chain dietary saturated fatty acid found in many animal and vegetable fats and oils. Deuteration, the incorporation of stable heavy isotopes into drug molecules, has garnered attention for its potential to influence pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-13572