Learn More
Medchemexpress LLC Stearic acid-d7 | 951209-59-9 | 98.0% | 291.52 g/mol | C18H29D7O2 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYB2219S910MG
Encompass_Preferred
Stearic acid-d7 is the deuterium-labeled analogue of stearic acid used as an isotopic internal standard and tracer in mass spectrometry and metabolic studies. It is chemically identical to native stearic acid but contains seven deuterium atoms, allowing mass differentiation while preserving chromatographic behavior. Supplied at high purity for analytical applications.
  • Deuterium-labeled (d7) stearic acid
  • High purity (98.0%)
  • Molecular weight 291.52 g/mol
  • Suitable as an internal standard for LC-MS/MS and GC-MS quantitation
  • Provided as a 10 MG analytical pack
Catalog No. 50-002-97483