Learn More
Cayman Chemical StearIc AcId glycIdyl STRd 1mg
SDP
Supplier:  Cayman Chemical 368391 MG
Encompass_Preferred
An internal standard for the quantification of stearic acid glycidyl ester by GC- or LC-MS
Catalog No. 50-290-9378