Learn More
Medchemexpress LLC Stearyl gallate | 10361-12-3 | 100 G

Supplier: Medchemexpress LLC HYN8082100G
Stearyl gallate is an alkyl gallate characterized by a long alkyl chain consisting of 18 carbon atoms. It exhibits antioxidant activity and a weak antiviral activity against HSV-1. For research use only.
- Alkyl gallate with an 18-carbon alkyl chain.
- Has antioxidant activity.
- Possesses weak antiviral activity against HSV-1.
- Purity: 99.50%.
- Appearance: Solid, white to off-white.
- Molecular weight: 422.60.
- Formula: C25H42O5.
- Soluble in DMSO (116.67 mg/mL or 276.08 mM).
- Potential applications in research related to antiviral and antioxidant studies.
- Available as a solid or in pre-made DMSO solutions for convenience.
- Detailed documentation including Data Sheet, COA, SDS, HNMR, LCMS, and Handling Instructions are provided.
- Can be used in various concentrations by preparing stock solutions.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.