Learn More
Medchemexpress LLC Stearyl gallate | 10361-12-3 | 25 G
SDP
Supplier:  Medchemexpress LLC 25GHYN8082
Encompass_Preferred
Stearyl gallate is an alkyl gallate featuring a long 18-carbon alkyl chain. This compound exhibits antioxidant activity and a weak antiviral activity against Herpes Simplex Virus type 1 (HSV-1). It is supplied as a white to off-white solid and is intended for research use only.
  • Exhibits antioxidant activity
  • Shows weak antiviral activity against HSV-1
  • High purity of 99.50%
  • Soluble in DMSO (116.67 mg/mL)
Catalog No. 50-004-21930