Learn More
Medchemexpress LLC Stearyl Stearate 1kg
SDP
Supplier:  Medchemexpress LLC HYW1274371KG
Encompass_Preferred
Stearyl StearateIt is an organic compound belonging to esters It is formed from the reaction between stearyl alcohol and stearic acid and has a mild fatty odor Stearyl stearate has several applications in the cosmetic and personal care industries especially as an emollient and conditioning agent in skin and hair care products Additionally it is used as a lubricant and surfactant in industrial settings
Catalog No. 50-002-06798