Learn More
Electron Microscopy Sciences Steel Ruler
SDP
Supplier:  Electron Microscopy Sciences 72909
Encompass_Preferred
Stainless steel ruler. 1 millimeter thick, 1 centimeter wide, 1 decimeter long. Its volume is 1 cubic centimeter and its weight in grams is the specific gravity. One edge is graduated in millimeters.
Catalog No. 50-291-22