Learn More
METASPEC Steel test panel for ASTM D1748 humidity cabinet testing. Each 2-in x 4-in test panel is polished and packaged individually. Sold in multiples of 10.
Supplier:  METASPEC 101A
Encompass
Steel test panels for use in the ASTM D1748 Humidity Cabinet test. This test measures the corrosion prevention or rust protection qualities of metal preservatives and rust inhibitors. ASTM D1748 is widely used by commercial, R&D, and military test laboratories.
These 1010 (ASTM A109) steel test panels are 2" x 4" x 1/8". The standard test panel has two 1/8" corner holes on one long side. The edges are rounded and the surface faces are finished to 10-20 rms micro "
Catalog No. NC3959116