Learn More
LGC GENOMICS LLC RNA FISH Hybridization Buffer, 10 mL
Supplier:  LGC GENOMICS LLC SMFHB110
Encompass_Preferred
RNA FISH Buffers contain proprietary additives to enhance detection with Stellaris RNA FISH probe sets, particularly for applications that typically exhibit more pronounced background fluorescence.
Catalog No. NC1242144