Learn More
iXCells Biotechnologies Stellate Cell Growth Medium
Supplier:  iXCells Biotechnologies MD0014500ML/DUP
Encompass_Preferred
Stellate Cell Growth Medium is designed to promote stellate cell proliferation in culture. This medium kit consists of Stellate Cell Basal Medium, 50 mL of Fetal Bovine Serum, 1 mL of Stellate Cell Growth Supplement, and 5 mL of Antibiotic ¿ Antimycotic (100X).
Catalog No. NC1467863