Learn More
Medchemexpress LLC Stem bromelain (EC 3.4.22.32) | 37189-34-7 | 25 G
SDP
Supplier:  Medchemexpress LLC 25GHYP2970
Encompass_Preferred
Stem bromelain (EC 3.4.22.32) is a cysteine protease and antibacterial agent isolated from pineapple (Ananas comosus), known for its fibrinolytic, anti-inflammatory, antitumor, and antimycobacterial activities.
  • Cysteine protease and antibacterial agent.
  • Induces secretion of IL-12p70 and IL-6.
  • Promotes apoptosis, affects PARP, Caspase 3, 9, Bax, and Bcl-2.
  • Fibrinolytic, antiedema, antithrombotic, and anti-inflammatory activities.
  • Shows in vivo antitumor, antileukemic, and antimetastatic effects.
  • Has antimycobacterial activity.
  • Provides protection against lead poisoning.
  • In vitro, causes apoptosis; increases CD80, CD86, IL-12p70, TNF-α, IL-6 in macrophages.
  • In vivo, reduces lead accumulation in kidneys and spleen of lead-exposed Wistar rats.
Catalog No. 50-003-96739