Learn More
Amsbio LLC STEM-CELLBANKER - GMP Grade (100ml)
Supplier:  Amsbio LLC 11924/DEL
Encompass
STEM-CELLBANKERGMP grade is a chemically defined, animal-free freezing medium manufactured under GMP conditions- optimized for ES cell and iPS cell storage as well as other valuable cell types. A Drug Master File (DMF) for STEM-CELLBANKER GMP grade has been registered by CBER (Center for Biologics Evaluation and Research) within the Food and Drug Administration.
Catalog No. NC0400932