Learn More
Amsbio LLC STEM-CELLBANKER - GMP Grade (20ml)
Supplier:  Amsbio LLC 11922/DEL
Encompass
STEM-CELLBANKERGMP grade is a chemically defined, animal-free freezing medium manufactured under GMP conditions- optimized for ES cell and iPS cell storage as well as other valuable cell types. A Drug Master File (DMF) for STEM-CELLBANKER GMP grade has been registered by CBER (Center for Biologics Evaluation and Research) within the Food and Drug Administration.
Catalog No. NC0261714
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.
View Alternate Product