Learn More
Amsbio LLC STEM-CELLBANKER DMSO FREE - GMP Grade (100ml)
Supplier:  Amsbio LLC 13926/DEL
Encompass
STEM-CELLBANKERGMP grade is a chemically defined, animal-free freezing medium manufactured under GMP conditions - optimized for ES cell and iPS cell storage as well as other valuable cell types.
Catalog No. NC1107357