Learn More
Medchemexpress LLC Stemazole 1mg | 317337-07-8 | 1 MG

Supplier: Medchemexpress LLC HY1160571MG
Stemazole is a small-molecule stem cell survival protective agent that promotes the survival and proliferation of human embryonic stem cells. It is supplied as a high-purity research reagent suitable for cellular assays and in vitro studies.
- Promotes stem cell survival and proliferation.
- High purity: 99.96%.
- Chemical formula C9H9N5OS2; molecular weight 267.34 g/mol.
- Provided as a solid suitable for preparation of stock solutions.
- Storage: store long-term at -20°C under inert gas; in solution, keep at -80°C for up to 6 months or -20°C for up to 1 month.
- Intended for research use in stem cell and cellular assays.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.