Learn More
Reprocell STEMOLECULE THIAZOVIVIN 1MG
Supplier:  Reprocell 040017
Encompass
Stemolecule™ Thiazovivin, 1Mg Thiazovivin Is A Selective, Cell Permeable Small Molecule That Directly Targets Rho-Associated Kinase (Rock). In Addition, Thiazovivin Protects Human Embryonic Stem Cells (Hescs) In The Absence Of Ecm By Regulating E-Cadherin Mediated Cell-Cell Interaction1. This Observation Suggests That Thiazovivin Promotes Cell Survival. In Other Studies Thiazovivin, In Combination With Inhibitors Of The Tgf-Β Receptor And Mek Pathway, Has Shown To Improve Reprogramming Efficiency By More Than 200-Fold. Purity: Greater Than 98% By Hplc Analysis Formulation: Light Brown Powder
Catalog No. NC0275323