Learn More
Apexbio Technology LLC StemRegenin 1 (SR1), 50mg. Cas: 1227633-49-9 MFCD: MFCD16987861. AhR antagonist
SDP
Supplier:  Apexbio Technology LLC A822450
Encompass_Preferred
StemRegenin 1 (SR1), a purine derivative, is a potent small molecule inhibitor of aryl hydrocarbon receptor (AhR) that potently antagonizes AhR with a value of 50% inhibition concentration IC50 of 127 nM in CD34+ cells. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5082