Learn More
Worthington Biochemical Corporation STEMXYME 1 COLLAGENAS 5X50MG
SDP
Supplier:  Worthington Biochemical Corporation LS004107
Encompass
A specialized combination of Animal Origin-Free Clostridium histolyticum collagenase (Code: CLSAFB) and animal origin free Bacillus polymyxa neutral protease with a minimum of 250 CLS units and 1,000 caseinase units per mg dry weight. Designed for stem cell and other primary cell isolations and bioprocessing applications where introduction of potential animal derived pathogens must be prevented. 2593-92-3
Catalog No. 50-592-485