Learn More
Grainger Step Up/Down Voltage Converter 500VA
Supplier:  Grainger 16V986
Encompass_Preferred
The Dayton Step-Up/Step-Down Voltage Transformer is a 120VAC input and output transformer that allows you to raise or lower voltage from 90 to 130VAC. It has a 500VA power capacity and includes a voltmeter to display output voltage. This transformer with adjustable voltage output is ideal for operating equipment when the supply voltage is higher or lower than required. It can be mounted vertically or horizontally.
Catalog No. 50-276-6282