Learn More
Medchemexpress LLC Stepronin (Prostenoglycine) | 72324-18-6 | 99.5% | C10H11NO4S2 | 100 MG
SDP
Supplier:  Medchemexpress LLC HYA0234100MG
Encompass_Preferred
Stepronin (Prostenoglycine) is an orally active expectorant, though inhalation administration is preferred over oral administration. It functions by inhibiting airway secretion in vitro, specifically by reducing Cl- secretion from epithelial cells and mucus glycoprotein secretion from submucal glands.
  • Orally active expectorant
  • Inhibits airway secretion in vitro
  • Reduces Cl- secretion from epithelial cells
  • Reduces mucus glycoprotein secretion from submucal glands
Catalog No. 50-004-32403