Learn More
Instech STER LOCK SOLN HEP/DEX 500USP 10ML
SDP
Supplier:  Instech USPHDS50010VY5
Encompass
Pharmaceutical-grade catheter lock solution prepared with heparin sodium following the USP <797> standard for compounding sterile preparations. The solution contains heparin (500 USP units/ml) in 50% dextrose, five 10ml vials, yellow.
Catalog No. NC2816895