Learn More
Cayman Chemical StercobIlInhydrochlorIde 5mg
SDP
Supplier:  Cayman Chemical 365095 MG
Encompass_Preferred
A fecal pigment and metabolite of bilirubin; induces DNA damage in a comet assay in HepG2 cells at 0.5-17 µM; induces activation of NF-κB in a reporter assay and increases the expression of the genes encoding TNF-α, IL-1β, IL-6, iNOS, and COX-2 in RAW 264.7 macrophages at 20 and 30 µM; fecal and plasma levels are increased in ob/ob mice
Catalog No. 50-291-2195