Learn More
Medchemexpress LLC Sterebin E | 114343-74-7 | 5 MG

Supplier: Medchemexpress LLC HYN81085MG
Sterebin E is a labdane diterpenoid isolated from *Stevia rebaudiana* leaves. This compound is intended for research purposes only and is not for patient use.
- Labdane diterpenoid
- Isolated from *Stevia rebaudiana* leaves
- Purity of 96.54%
- Molecular weight: 338.48
- Molecular formula: C20H34O4
- Appearance: Solid
- Color: Off-white to light yellow
- Classified as terpenoids, diterpenoids
- Initial source: Plants
- Shipping conditions: Room temperature
- Storage for powder: -20°C for 3 years
- Storage in solvent: -80°C for 6 months, -20°C for 1 month
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.