Learn More
Cayman Chemical SterIgmatocystIn 10mg
SDP
Supplier:  Cayman Chemical 1144110 MG
Encompass_Preferred
A mycotoxin; lethal to rats (LD50 = 60 mg/kg) and chick embryos (LD50s = 5-7 UG/embryo); induces gastritis, as well as intestinal polyp formation and metaplasia in aged Mongolian gerbils at 100 and 1,000 ppb in drinking water; induces formation of lung and liver adenomas and malignant lymphomas in newborn mice
Catalog No. 50-295-2422