Learn More
Cayman Chemical SterIgmatocystIn-13C18 1.2mL
SDP
Supplier:  Cayman Chemical 313021.2 ML
Encompass_Preferred
An internal standard for the quantification of sterigmatocystin by GC- or LC-MS
Catalog No. 50-298-8425