Learn More
Medchemexpress LLC HY-N6725 5mg Medchemexpress, Sterigmatocystine CAS:10048-13-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN67255MG
Encompass_Preferred
Medchemexpress, HY-N6725 5mg Sterigmatocystine CAS:10048-13-2 Sterigmatocystine is a precursor of aflatoxins and a mycotoxin produced by common mold strains from Aspergillus versicolor. Sterigmatocystine, a inhibitor of G1 Phase and DNA synthesis, is used to inhibit p21 activity. Sterigmatocystine has teratogenic, and carcinogenic effects in animals. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2958