Learn More
Bellco Glass, Inc. Sterile Forcep 128mm Blue
SDP
Supplier:  Bellco Glass, Inc. 130000382
Encompass_Preferred
Forceps are self-sprung and sturdy enough to be reusable, but economical enough to be disposable.Available in:Acetal (Green) - Resistant to solvents, acid and alkaliABS (Blue) - Features pointed end. Available in bulk packs and in individual sterile packsPolystyrene (Transparent Blue) - Available in bulk packs and in individual sterile packs. Type-Sterile, Length (mm)-130, Material-ABS, Color-Blue, Package Type-Individual pack, Carton Qty-100
Catalog No. 50-233-0720